Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corin Antibody (MM0199-9H21) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
| Antigen | Corin |
|---|---|
| Clone | MM0199-9H21 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Corin Monoclonal specifically detects Corin in Human samples. It is validated for Western Blot.Specifications
| Corin | |
| LYOPH | |
| Unconjugated | |
| Rat | |
| Human | |
| atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10 | |
| CORIN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0199-9H21 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9Y5Q5 | |
| 10699 | |
| IgG2 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title