Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCIR/CLEC4A Antibody (MM0237-5P20) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
| Antigen | DCIR/CLEC4A |
|---|---|
| Clone | MM0237-5P20 |
| Concentration | LYOPH |
| Applications | CyTOF, Western Blot, Flow Cytometry |
| Classification | Monoclonal |
Description
DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| DCIR/CLEC4A | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor | |
| CLEC4A | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0237-5P20 | |
| CyTOF, Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity | |
| Q9UMR7 | |
| 50856 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title