Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax2 Antibody (PAX2/1104), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB011643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011643 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011643 Supplier Novus Biologicals Supplier No. NBP279877JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pax2 Monoclonal specifically detects Pax2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Pax2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PAX2/1104
Conjugate Janelia Fluor 646
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2
Gene Symbols PAX2
Host Species Mouse
Immunogen Recombinant fragment (around aa 223-354) human Pax2 protein (exact sequence is proprietary) (Uniprot: Q02962)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Test Specificity This antibody recognizes a protein of 42kDa, which is identified as Pax2. Pax2 has been used as a marker for the identification of renal cell carcinoma and ovarian carcinoma by immunohistochemistry.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.