Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax3 Antibody (HL160), Novus Biologicals™
SDP

Catalog No. NB177348 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177348 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177348 Supplier Novus Biologicals Supplier No. NBP313661
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Pax3 Monoclonal antibody specifically detects Pax3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Pax3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL160
Conjugate Unconjugated
Formulation PBS
Gene Alias CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3
Host Species Rabbit
Immunogen The immunogen used to generate this antibody corresponds to Coturnix coturnix Pax3.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5077
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.