Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax3 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001420 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001420 Supplier Novus Biologicals Supplier No. NB1001420
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 3 publications

Pax3 Polyclonal specifically detects Pax3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Peptide ELISA, Western Blot.

Specifications

Antigen Pax3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 3 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Peptide ELISA detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000429
Gene Alias CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3
Gene Symbols PAX3
Host Species Goat
Immunogen Peptide with sequence TTLAGAVPRMM-C corresponding to N-Terminus according to NP_039230.1, NP_852122.1, NP_852123.1, NP_852124.1, NP_852125.1, NP_852126.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5077
Test Specificity This antibody is expected to recognize all 7 reported human isoforms (as represented by NP_000429.2; NP_039230.1; NP_852122.1;.1 NP_852123.1; NP_852124.1; NP_852125.1 and NP_852126.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.