Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax4 Antibody (3C12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237644
Description
Pax4 Monoclonal specifically detects Pax4 in Human samples. It is validated for Western Blot, ELISA.Specifications
Pax4 | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
PAX4 | |
Purified recombinant fragment of human PAX4 expressed in E. Coli. | |
Unpurified | |
RUO | |
5078 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA | |
3C12 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4 | |
Mouse | |
38 kDa | |
0.1 mL | |
Primary | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction