Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax4 Antibody (3C12), Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | Pax4 |
---|---|
Clone | 3C12 |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Pax4 Monoclonal specifically detects Pax4 in Human samples. It is validated for Western Blot, ELISA.Specifications
Pax4 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Ascites | |
RUO | |
Ascites with 0.03% Sodium Azide | |
5078 | |
Purified recombinant fragment of human PAX4 expressed in E. Coli. | |
Primary | |
38 kDa |
3C12 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4 | |
PAX4 | |
IgG1 | |
Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title