Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax4 Antibody (OTI2F3), FITC, Novus Biologicals™
SDP

Catalog No. NB302069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302069 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302069 Supplier Novus Biologicals Supplier No. NBP273257F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pax4 Monoclonal antibody specifically detects Pax4 in Human samples. It is validated for Western Blot

Specifications

Antigen Pax4
Applications Western Blot
Classification Monoclonal
Clone OTI2F3
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-228 of human PAX4 (NP_006184) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5078
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.