Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax5/BSAP Antibody (PAX5/2060), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB383913 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383913 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383913 Supplier Novus Biologicals Supplier No. NBP308873JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Pax5/BSAP
Applications Western Blot, ELISA
Classification Monoclonal
Clone PAX5/2060
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
Host Species Mouse
Immunogen Recombinant full-length human Pax5/BSAP protein (Uniprot: Q02548)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5079
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.