Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax6 Antibody, Novus Biologicals™
SDP

Catalog No. NB10061654 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061654 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061654 Supplier Novus Biologicals Supplier No. NB10061654
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Pax6 Polyclonal specifically detects Pax6 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen) (Negative), Peptide ELISA, Western Blot.

Specifications

Antigen Pax6
Applications Flow Cytometry, Immunohistochemistry (Frozen) (Negative), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000, Immunohistochemistry-Frozen (Negative)
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000271.1
Gene Alias keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6
Gene Symbols PAX6
Host Species Goat
Immunogen Peptide with sequence C-REEKLRNQRRQASN corresponding to internal region according to NP_001595.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Diabetes Research, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5080
Test Specificity This antibody is expected to recognize both reported isoforms (NP_000271.1 and NP_001595.2).
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.