Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax6 Antibody (PAX6/7078R), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB178452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB178452 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB178452 Supplier Novus Biologicals Supplier No. NBP314033AF647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Pax6 Monoclonal antibody specifically detects Pax6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence

Specifications

Antigen Pax6
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PAX6/7078R
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6
Host Species Rabbit
Immunogen Recombinant fragment (N-terminus; aa 1-300) of human Pax6 protein (Uniprot: P26367)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5080
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.