Learn More
Description
Specifications
Specifications
| Antigen | Pax6 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PAX6/7078R |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 |
| Host Species | Rabbit |
| Immunogen | Recombinant fragment (N-terminus; aa 1-300) of human Pax6 protein (Uniprot: P26367) |
| Purification Method | Protein A or G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
