Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax7 Antibody (PAX7/1187), Novus Biologicals™
SDP

Catalog No. NBP24458802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24458802 0.2 mg
NBP24458800 0.02 mg
NBP24458801 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24458802 Supplier Novus Biologicals Supplier No. NBP2445880.2MG

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Immunohistochemistry (Paraffin)

Pax7 Monoclonal antibody specifically detects Pax7 in Human samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen Pax7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PAX7/1187
Concentration 0.2mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P23759
Gene Alias FLJ37460, HuP1, paired box 7, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7, PAX7 transcriptional factor, PAX7B, RMS2
Gene Symbols PAX7
Host Species Mouse
Immunogen Recombinant fragment (aa300-600) of human PAX7 protein
Molecular Weight of Antigen 57 kDa
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5081
Test Specificity The Pax gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the Pax family of proteins have been linked to disease and cancer in humans. Pax-7 is a protein specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization reveals that Pax-7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding Pax-7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma).
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.