Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX8 Antibody (ZR-1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB381728 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381728 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381728 Supplier Novus Biologicals Supplier No. NBP308307R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen PAX8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZR-1
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of human PAX8 protein (Uniprot: Q06710)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7849
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.