Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paxillin Antibody (GT722), Novus Biologicals™
SDP

Catalog No. NB177249 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177249 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177249 Supplier Novus Biologicals Supplier No. NBP313562
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Paxillin Monoclonal antibody specifically detects Paxillin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Paxillin
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone GT722
Conjugate Unconjugated
Dilution Western Blot, Immunofluorescence 1:100-1:1000
Formulation PBS
Gene Alias FLJ16691, paxillin
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide corresponding to residues around human Paxillin (non-phospho Tyr118). The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5829
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.