Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paxillin Antibody (LD4 sAB), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB015197 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015197 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015197 Supplier Novus Biologicals Supplier No. NBP281063G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Paxillin Monoclonal specifically detects Paxillin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Paxillin
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone LD4 sAB
Conjugate DyLight 488
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ16691, paxillin
Gene Symbols PXN
Host Species Rabbit
Immunogen Peptide corresponding to human paxillin-LD4 motif (residues 260-278) fused to MBP; peptide sequence: WGGSATRELDELMASLSD.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5829
Test Specificity The antibody binds specifically to the LD-4 motif of human Paxillin with a Kd of 6 nM.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.