Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paxillin Rabbit anti-Human, Mouse, Rat, Clone: 2E6P0, Novus Biologicals™
SDP

Catalog No. NB168937 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168937 100 μg
NB168938 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168937 Supplier Novus Biologicals Supplier No. NBP316929100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Paxillin Monoclonal antibody specifically detects Paxillin in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Paxillin
Applications Western Blot
Classification Monoclonal
Clone 2E6P0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FLJ16691, paxillin
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around Y118 of human Paxillin (P49023).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5829
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.