Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PB 28 dihydrochloride
GREENER_CHOICE

Catalog No. p-7112876
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity σ2 agonist

PB 28 dihydrochloride is a high affinity σ2 receptor agonist (Ki values are 0.8 and 15.2 nM for σ2 and σ1 receptors respectively) that displays minimal affinity at other receptors. Inhibits electrically evoked twitch in guinea pig bladder and ileum (EC50 values are 2.62 and 3.96 μM respectively). Displays antiproliferative and cytotoxic effects in SK-N-SH neuroblastoma and C6 glioma cells.

Specifications

Chemical Name or Material 1-Cyclohexyl-4-[3-(1,2,3,4-tetrahydro-5-methoxy-1-naphthalenyl)propyl]piperazine dihydrochloride
CAS 172907-03-8
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 20mM with gentle warming in water
Molecular Formula C24H38N2O.2HCl
Formula Weight Observed MW: 443.5

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.