Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBEF/Visfatin/NAMPT Antibody (OMNI379), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB172272 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172272 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172272 Supplier Novus Biologicals Supplier No. NBP280036G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PBEF/Visfatin/NAMPT Monoclonal antibody specifically detects PBEF/Visfatin/NAMPT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PBEF/Visfatin/NAMPT
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OMNI379
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN
Host Species Mouse
Immunogen Recombinant human PBEF/Visfatin/NAMPT (visfatin/PBEF).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10135
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.