Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBR Antibody, Novus Biologicals™
SDP

Catalog No. NB10041398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10041398 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10041398 Supplier Novus Biologicals Supplier No. NB10041398
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PBR Polyclonal specifically detects PBR in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Peptide ELISA, Western Blot.

Specifications

Antigen PBR
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-0.3 ug/ml, Flow Cytometry 10 ug/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 2-4 ug/ml, Peptide ELISA detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P30536
Gene Alias benzodiazepine peripheral binding site, BZRPDBI, IBP, MBRPBS, mDRC, Mitochondrial benzodiazepine receptor, PBRBPBS, PBRS, peripheral benzodiazepine receptor, Peripheral-type benzodiazepine receptor, pk18, PKBSbenzodiazapine receptor (peripheral), PTBR, translocator protein, translocator protein (18kDa)
Gene Symbols TSPO
Host Species Goat
Immunogen Peptide with sequence C-RDNHGWRGGRRLPE corresponding to C-Terminus according to NP_000705.2.
Molecular Weight of Antigen 18.8 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 706
Test Specificity This antibody is expected to recognize one isoform (NP_000705.2) only.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.