Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBX1 Antibody (PCRP-PBX1-3C8), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB178505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB178505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB178505 Supplier Novus Biologicals Supplier No. NBP314035FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PBX1 Monoclonal antibody specifically detects PBX1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen PBX1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-PBX1-3C8
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias DKFZp686B09108, Homeobox protein PBX1, Homeobox protein PRL, MGC126627, Pre-B cell leukemia transcription factor-1, pre-B-cell leukemia homeobox 1, pre-B-cell leukemia transcription factor 1, PRL
Host Species Mouse
Immunogen Recombinant full-length human PBX1 protein (Uniprot: P40424)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5087
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.