Learn More
Pc2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | Pc2 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2504
![]() |
Bethyl Laboratories, Inc
IHC00668T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2503
![]() |
Bethyl Laboratories, Inc
IHC00668 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Rabbit, Guinea pig_10141, Pig, Panda, Monkey, Gorilla, Chimpanzee, Gray short
CBX4 is a member of Drosophila Polycomb group gene family. The polycomb group (PcG) genes are essential for maintece of proper expression patterns of developmental master regulators; changes in expression pf PcG proteins have been associated with cancer. CBX4 is a part of the cellular memory system responsible for the inheritance of gene activity by progeny cells. It participates in maintaining the transcriptionally repressive state of genes. CBX4 is part of acomplex that acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. CBX4 is an E3 SUMO-protein ligase which facilitates SUMO1 conjugation by UBE2I.Specifications
Pc2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
E3 SUMO-protein ligase CBX4; Chromobox protein homolog 4; Polycomb 2 homolog; Pc2; hPc2 | |
Between 150 and 200 | |
Primary | |
4° C | |
CBX4 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O00257, O55187 | |
12418, 8535 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.