Learn More
Description
Ensure accurate, reproducible results in Western Blot, ELISA
Specifications
Specifications
| Antigen | PCBP1 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 2A10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | NP_006187 |
| Gene Alias | alpha-CP1, Heterogeneous nuclear ribonucleoprotein E1, heterogenous nuclear ribonucleoprotein E1, heterogenous nuclear ribonucleoprotein X, hnRNP E1, hnRNP-E1, hnRNP-X, HNRPE1, HNRPX, nucleic acid binding protein sub 2.3, Nucleic acid-binding protein SUB2.3, poly(rC) binding protein 1, poly(rC)-binding protein 1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
