Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCCA Antibody, Novus Biologicals™
SDP

Catalog No. NBP219717 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219717 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219717 Supplier Novus Biologicals Supplier No. NBP219717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCCA Polyclonal specifically detects PCCA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PCCA
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P05165
Gene Alias EC 6.4.1, EC 6.4.1.3, pccA complementation group, PCCase alpha subunit, PCCase subunit alpha, propanoyl-CoA:carbon dioxide ligase alpha subunit, Propanoyl-CoA:carbon dioxide ligase subunit alpha, propionyl CoA carboxylase, alpha polypeptide, propionyl Coenzyme A carboxylase, alpha polypeptide, propionyl-CoA carboxylase alpha chain, mitochondrial
Gene Symbols PCCA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PCCA. The exact sequence is proprietary.
Molecular Weight of Antigen 80 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5095
Test Specificity Species reactivity based on sequence identity: Mouse (86%), Rat (86%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.