Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDH11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179198
Description
PCDH11 Polyclonal specifically detects PCDH11 in Mouse samples. It is validated for Western Blot.Specifications
PCDH11 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
PCDH11, PCDH11Y, PCDH22, PCDH-PC, PCDHX, PCDH-X, PCDHY, PCDH-Y, protein phosphatase 1, regulatory subunit 119, protocadherin 11X, protocadherin 22, Protocadherin on the X chromosome, Protocadherin on the Y chromosome, Protocadherin prostate cancer, protocadherin-11 X-linked, Protocadherin-11 Y-linked, Protocadherin-22, protocadherin-PC, Protocadherin-S | |
Rabbit | |
Affinity purified | |
RUO | |
27328 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_001074854 | |
PCDH11 | |
The immunogen for this antibody is Pcdh11x. Peptide sequence LNQSSMLLIKVKDENDNAPVFTQSFISLSVPENNSPGAQLTKISATDADS. | |
100 μL | |
Primary | |
The immunogen sequence has 100% identity to both PCDH11X and PCDH11Y. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction