Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHA5 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB132425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132425 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132425 Supplier Novus Biologicals Supplier No. NBP306113H

Rabbit Polyclonal Antibody

PCDHA5 Polyclonal specifically detects PCDHA5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PCDHA5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CNR6, CNRN6, CNRS6, CRNR6, KIAA0345-like 9, ortholog of mouse CNR6, PCDH-alpha-5, PCDH-ALPHA5, protocadherin alpha 5, protocadherin alpha-5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PCDHA5 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56143
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.