Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHGA1 Antibody (S159-5), HRP, Novus Biologicals™
SDP

Catalog No. NBP242207H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242207H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242207H Supplier Novus Biologicals Supplier No. NBP242207H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PCDHGA1 Monoclonal specifically detects PCDHGA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PCDHGA1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone S159-5
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias MGC138287, MGC138289, PCDH-GAMMA-A1, protocadherin gamma subfamily A, 1, protocadherin gamma-A1
Gene Symbols PCDHGA1
Host Species Mouse
Immunogen Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ∼807-930, B subfamily amino acids ∼789-912 and C subfamily amino acids ∼818-941). 99% identity with human (123/124 amino acids).
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 56114
Test Specificity Detects approx 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.