Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHGC5 Antibody (OTI2F3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB302057 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302057 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302057 Supplier Novus Biologicals Supplier No. NBP273270APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PCDHGC5 Monoclonal antibody specifically detects PCDHGC5 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PCDHGC5
Applications Western Blot
Classification Monoclonal
Clone OTI2F3
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias MGC138286, MGC138288, PCDH-GAMMA-C5, protocadherin gamma subfamily C, 5, protocadherin gamma-C5
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 35-258 of human PCDHGC5(NP_115783) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56097
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.