Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCF11 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB120154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120154 25 μg
NB120153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB120154 Supplier Novus Biologicals Supplier No. NBP31100325UL

Rabbit Polyclonal Antibody

PCF11 Polyclonal specifically detects PCF11 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen PCF11
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias KIAA0824PCF11p homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog (S. cerevisiae), PCF11, cleavage and polyadenylation factor subunit, homolog (S. cerevisiae), pre-mRNA cleavage complex 2 protein Pcf11, Pre-mRNA cleavage complex II protein Pcf11
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the following sequence SSEHRLAGSRNKIINGIVQKQDTITEESEKQGTKPGRSSTRKRSRSRSPK (NP_056969). Peptide sequence SSEHRLAGSRNKIINGIVQKQDTITEESEKQGTKPGRSSTRKRSRSRSPK
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51585
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.