Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTN1-C Antibody (RNL-4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB003013 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003013 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003013 Supplier Novus Biologicals Supplier No. NBP278849
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RTN1-C Monoclonal antibody specifically detects RTN1-C in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen RTN1-C
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RNL-4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10 - 1:25, Immunohistochemistry 1:10 - 1:25, Immunohistochemistry-Paraffin 1:10 - 1:25, Immunohistochemistry-Frozen 1:10 - 1:25
Formulation PBS with 0.09% Sodium Azide
Gene Alias Neuroendocrine-specific protein, NSP, reticulon-1, reticulon-1C
Gene Symbols RTN1
Host Species Mouse
Immunogen Synthetic peptide encompassing the unique 20 N-terminal amino acid sequence of RTN1-C
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6252
Test Specificity Recognizing epitopes in the neuro-endocrine specific proteins (NSP) C. This antibody was raised against the amino-terminal amino acid sequence of NSP-C. cross reactivity is observed with human and rat.
Target Species Human, Rat
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.