Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCIF1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB046490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB046490 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB046490 Supplier Novus Biologicals Supplier No. NBP297683AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCIF1 Polyclonal specifically detects PCIF1 in Human samples. It is validated for Western Blot.

Specifications

Antigen PCIF1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias bA465L10.1, C20orf67, chromosome 20 open reading frame 67, PDX-1 C terminus-interacting factor 1, PDX1 C-terminal inhibiting factor 1, phosphorylated CTD-interacting factor 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PCIF1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63935
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.