Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCIF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112227 Supplier Invitrogen™ Supplier No. PA5112227
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCIF1 Polyclonal Antibody for Western Blot, IP

PCIF1 may play a role in transcription elongation or in coupling transcription to pre-mRNA processing through its association with the phosphorylated C-terminal domain (CTD) of RNAPII largest subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCIF1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene PCIF1
Gene Accession No. Q9H4Z3
Gene Alias 2310022K11Rik; C20orf67; CAPAM; Cap-specific adenosine methyltransferase; F730014I05Rik; hPCIF1; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; Pcif1; PDX-1 C terminus-interacting factor 1; PDX1 C-terminal inhibiting factor 1; phosphorylated CTD interacting factor PCIF1; phosphorylated CTD-interacting factor 1; PPP1R121; protein phosphatase 1, regulatory subunit 121; RGD1310800
Gene Symbols PCIF1
Host Species Rabbit
Immunogen E. coli-derived Human PCIF1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63935
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.