Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCK2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219729 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219729 Supplier Novus Biologicals Supplier No. NBP219729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCK2 Polyclonal specifically detects PCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PCK2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q16822
Gene Alias EC 4.1.1.32, PE, PEPCK, PEPCK2PEP carboxykinase, PEPCK-M, phosphoenolpyruvate carboxykinase [GTP], mitochondrial, phosphoenolpyruvate carboxykinase 2 (mitochondrial), Phosphoenolpyruvate carboxylase, phosphopyruvate carboxylase
Gene Symbols PCK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human PCK2. The exact sequence is proprietary.
Molecular Weight of Antigen 71 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5106
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.