Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCK2 Rabbit anti-Human, Mouse, Rat, Clone: 10L10J8, Novus Biologicals™
SDP

Catalog No. NB167927 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167927 100 μg
NB167928 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167927 Supplier Novus Biologicals Supplier No. NBP316418100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PCK2 Monoclonal antibody specifically detects PCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen PCK2
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 10L10J8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 4.1.1.32, PE, PEPCK, PEPCK2PEP carboxykinase, PEPCK-M, phosphoenolpyruvate carboxykinase [GTP], mitochondrial, phosphoenolpyruvate carboxykinase 2 (mitochondrial), Phosphoenolpyruvate carboxylase, phosphopyruvate carboxylase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 550-640 of human PEPCK/PCK2 (Q16822).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5106
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.