Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Antibody (144), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB037157 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037157 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037157 Supplier Novus Biologicals Supplier No. NBP290058AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PCNA Monoclonal specifically detects PCNA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PCNA
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 144
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PCNA (P12004; Met1-Ser261).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5111
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.