Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Antibody (5E6/2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20238A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20238A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20238A94 Supplier Novus Biologicals Supplier No. NBP250238AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PCNA Monoclonal specifically detects PCNA in Yeast samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PCNA
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5E6/2
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Gene Symbols PCNA
Host Species Mouse
Immunogen Purified recombinant His6-PCNA from budding yeast, Saccharomyces cerevisiae, produced in Escherichia coli
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5111
Target Species Yeast
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.