Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Antibody (PC10), Novus Biologicals™
SDP

Catalog No. NB500106SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500106SS 0.025 mL
NB500106 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500106SS Supplier Novus Biologicals Supplier No. NB500106SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 35 publications

PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Specifications

Antigen PCNA
Applications Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone PC10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Simple Western 1:200, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 26883631), Flow Cytometry 1:10-1:1000, ELISA reported in scientific literature, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000-1:2000. Use reported in scientific literature (PMID 22068968), Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 27212918), Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Gene Accession No. P12004
Gene Alias cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Gene Symbols PCNA
Host Species Mouse
Immunogen Protein A-rat PCNA fusion obtained from pC2T
Molecular Weight of Antigen 30 kDa
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cellular Markers, DNA Repair, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 5111
Test Specificity Specific for PCNA p36 protein expressed at high levels in proliferating cells.
Target Species Human, Mouse, Rat, Pig, Chicken, Drosophila, Fish, Marsupial, Primate, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.