Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Antibody (PC10), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB500106R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500106R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500106R Supplier Novus Biologicals Supplier No. NB500106R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PCNA Monoclonal specifically detects PCNA in Human, Mouse, Rat, Porcine, Chicken, Drosophila, Fish, Primate, Rabbit, Yeast, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen PCNA
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PC10
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. P12004
Gene Alias cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Gene Symbols PCNA
Host Species Mouse
Immunogen Protein A-rat PCNA fusion obtained from pC2T
Molecular Weight of Antigen 30 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5111
Test Specificity Specific for PCNA p36 protein expressed at high levels in proliferating cells.
Target Species Human, Mouse, Rat, Pig, Chicken, Drosophila, Fish, Primate, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.