Learn More
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
Specifications
| Antigen | PCNA |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCNA/694 |
| Concentration | 0.2mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Formulation | 10mM PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. | P12004 |
| Gene Alias | cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
