Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA associated factor Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290011 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290011 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290011 Supplier Novus Biologicals Supplier No. NBP232117APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCNA associated factor Polyclonal antibody specifically detects PCNA associated factor in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)

Specifications

Antigen PCNA associated factor
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative)
Formulation PBS
Gene Alias HCV NS5A-transactivated protein 9, Hepatitis C virus NS5A-transactivated protein 9, KIAA0101, NS5ATP9p15PAF, OEATC-1OEATC1, Overexpressed in anaplastic thyroid carcinoma 1, p15(PAF), PAF, PCNA-associated factor
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human PCNA-Associated Factor using the numbering given in entry NP_055551.1 (GeneID 9768).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9768
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.