Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA associated factor Antibody (K11-P1G7*B8), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP250536AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250536AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250536AG Supplier Novus Biologicals Supplier No. NBP250536AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PCNA associated factor Monoclonal specifically detects PCNA associated factor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen PCNA associated factor
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone K11-P1G7*B8
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HCV NS5A-transactivated protein 9, Hepatitis C virus NS5A-transactivated protein 9, KIAA0101, NS5ATP9p15PAF, OEATC-1OEATC1, Overexpressed in anaplastic thyroid carcinoma 1, p15(PAF), PAF, PCNA-associated factor
Gene Symbols PCLAF
Host Species Mouse
Immunogen Peptide Sequence - RKACPLQPDHT -peptide immunogen is identical in mouse (Mus musculus) and rat (Rattus norvegicus).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9768
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.