Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNP Antibody (OTI4E3), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N273272A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273272A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273272A647 Supplier Novus Biologicals Supplier No. NBP273272AF647

Mouse Monoclonal Antibody

PCNP Monoclonal specifically detects PCNP in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PCNP
Applications Western Blot
Classification Monoclonal
Clone OTI4E3
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp781I24156, PEST proteolytic signal containing nuclear protein, PEST proteolytic signal-containing nuclear protein, PEST-containing nuclear protein
Gene Symbols PCNP
Host Species Mouse
Immunogen Full length recombinant protein of human PCNP (NP_065090) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 57092.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.