Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNX3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP261232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261232 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261232 Supplier Novus Biologicals Supplier No. NBP261232

Rabbit Polyclonal Antibody

PCNXL3 Polyclonal specifically detects PCNXL3 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen PCNX3
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation 8 - 15 ul/mg lysate
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Symbols PCNX3
Host Species Rabbit
Immunogen between 1984 and 2034
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 399909
Target Species Human
Content And Storage Store the unopened product at 2 - 8degrees C. Protect from light. Do not use past expiration date.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.