Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCP4 Antibody (CL5306), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | PCP4 |
---|---|
Clone | CL5306 |
Dilution | Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PCP4 Monoclonal specifically detects PCP4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PCP4 | |
Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Unconjugated | |
Mouse | |
brain specific polypeptide PEP19, 10Brain-specific antigen PCP-4, Brain-specific polypeptide PEP-19, PEP19, PEP-19, Purkinje cell protein 4 | |
PCP4 | |
IgG1 | |
Protein A purified |
CL5306 | |
Monoclonal | |
Purified | |
Human, Mouse, Rat | |
5121 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GAGATNGKDKTSGENDGQKKVQEEFDIDMDAPETERAAVAIQSQFRKFQKKK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title