Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCP4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB187973 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187973 200 μg
NB187974 50 μg
NB301764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB187973 Supplier Novus Biologicals Supplier No. NBP314649200UL
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PCP4 Polyclonal antibody specifically detects PCP4 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen PCP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:100-1:500
Formulation PBS, pH 7.0 with 0.03% Proclin 300
Gene Alias brain specific polypeptide PEP19, 10Brain-specific antigen PCP-4, Brain-specific polypeptide PEP-19, PEP19, PEP-19, Purkinje cell protein 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PCP4 fragment,
Purification Method Antigen and protein A Affinity-purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5121
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.