Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCYOX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | PCYOX1 |
---|---|
Concentration | 0.88 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119353
|
Invitrogen™
PA5119353 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121698
|
Invitrogen™
PA5121698 |
20 μL |
Each for $209.50
|
|
|||||
Description
PCYOX1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Prenylcysteine is released during the degradation of prenylated proteins. PCYOX1 catalyzes the degradation of prenylcysteine to yield free cysteines and a hydrophobic isoprenoid product (Tschantz et al., 1999 ).Specifications
PCYOX1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
PCYOX1 | |
1200015P13Rik; AI115532; AI448340; chloride ion pump-associated 55 kDa protein; Clp55; KIAA0908; PCL1; Pcly; PCYOX1; Prenylcysteine lyase; prenylcysteine oxidase; Prenylcysteine oxidase 1; prenylcysteine oxidase; prenylcysteine oxidase 1; prenylcysteine oxidase-like; UNQ597/PRO1183 | |
PCYOX1 | |
Primary | |
Antigen affinity chromatography |
0.88 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9UHG3 | |
51449 | |
Recombinant protein Prenylcysteine Oxidase 1. The antigen corresponds to amino acid range 174-505 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title