Learn More
Description
Specifications
Specifications
| Antigen | PCYOX1L |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 1.8.3.-, MGC3265, prenylcysteine oxidase 1 like, prenylcysteine oxidase-like |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human PCYOX1L fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
