Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCYT1B Antibody (F39 P7 F11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP229807 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229807 0.1 mg
NB019470 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229807 Supplier Novus Biologicals Supplier No. NBP229807
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PCYT1B Monoclonal specifically detects PCYT1B in Human samples. It is validated for Western Blot.

Specifications

Antigen PCYT1B
Applications Western Blot
Classification Monoclonal
Clone F39 P7 F11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 01 - 05 ug/ml
Gene Accession No. Q9Y5K3
Gene Alias CCT B, CCTB, CCT-betaEC 2.7.7.15, choline-phosphate cytidylyltransferase B, CT B, CTB, CTP:phosphocholine cytidylyltransferase B, EC 2.7.7, phosphate cytidylyltransferase 1, choline, beta, phosphate cytidylyltransferase 1, choline, beta isoform, Phosphorylcholine transferase B
Gene Symbols PCYT1B
Host Species Mouse
Immunogen Synthetic Peptide corresponding to the C-Terminus of Human CCT-beta.
Molecular Weight of Antigen 60 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9468
Test Specificity CCT-beta.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.