Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Antibody (2335A) [FITC], Novus Biologicals™
SDP

Catalog No. NB401139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401139 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401139 Supplier Novus Biologicals Supplier No. FAB10863F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF

Specifications

Antigen PD-1
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 2335A
Conjugate FITC
Dilution Western Blot, Flow Cytometry, ELISA, CyTOF-ready
Formulation PBS
Gene Alias CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human PD-1, Met1-Gln167, Accession # Q15116
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.