Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Antibody (2335A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339902 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339902 Supplier Novus Biologicals Supplier No. FAB10863MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen PD-1
Applications CyTOF, ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2335A
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, ELISA, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human PD-1, Met1-Gln167, Accession # Q15116
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.